Learn More
RNA Polymerase II/POLR2A Antibody (POLR2A/9089R), Alexa Fluor™ 405, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-24041AF405
Product Code. 18514909
  • £514.00 / 0.10mL
Description
RNA Polymerase II/POLR2A Monoclonal antibody specifically detects RNA Polymerase II/POLR2A in Human, Yeast samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RNA Polymerase II/POLR2A
Monoclonal
Alexa Fluor 405
DNA-directed RNA polymerase II largest subunit, RNA polymerase II 220 kdsubunit, DNA-directed RNA polymerase II subunit A, DNA-directed RNA polymerase II subunit RPB1, DNA-directed RNA polymerase III largest subunit, EC 2.7.7.48, EC 2.7.7.6, hRPB220, hsRPB1, MGC75453, POLR2, POLRA, polymerase (RNA) II (DNA directed) polypeptide A (220kD), polymerase (RNA) II (DNA directed) polypeptide A, 220kDa, RNA Pol 2, RNA Polymerase 2, RNA polymerase II subunit B1, RNA-directed RNA polymerase II subunit RPB1, RNAPII, RPB1, RPBh1, RpIILS, RPO2, RPOL2
Ten repeats of synthetic peptide YSPTSPS using chemically synthesized phospho-Ser5
0.1 mL
Chromatin Research, Epigenetics, Phospho Specific, Transcription Factors and Regulators
5430
Store at 4°C in the dark.
IgG κ
Western Blot, Immunohistochemistry (Paraffin)
POLR2A/9089R
50 mM Sodium Borate
Rabbit
Protein A purified
RUO
Primary
Human, Yeast
Purified