missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNA Polymerase II/POLR2A Antibody (C.15200004), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59213
This item is not returnable.
View return policy
Description
RNA Polymerase II/POLR2A Monoclonal antibody specifically detects RNA Polymerase II/POLR2A in Human, Xenopus, Yeast, Zebrafish samples. It is validated for Western Blot, ChIP assay, ELISA, Immunocytochemistry, Immunofluorescence, Knockdown.
Specifications
| RNA Polymerase II/POLR2A | |
| Monoclonal | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| POLR2A | |
| The exact immunogen is propietary information. | |
| 50 μg | |
| Chromatin Research, Epigenetics, microRNA, Transcription Factors and Regulators | |
| 5430 | |
| Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, ChIP Assay | |
| C.15200004 | |
| Western Blot 1:1000, ELISA 1:3000, Immunocytochemistry/Immunofluorescence 1:500, Chromatin Immunoprecipitation (ChIP) 1 ug/IP, Chromatin Immunoprecipitation Sequencing 1ug/ChIP, Knockdown Validated | |
| DNA-directed RNA polymerase II largest subunit, RNA polymerase II 220 kdsubunit, DNA-directed RNA polymerase II subunit A, DNA-directed RNA polymerase II subunit RPB1, DNA-directed RNA polymerase III largest subunit, EC 2.7.7.48, EC 2.7.7.6, hRPB220, hsRPB1, MGC75453, POLR2, POLRA, polymerase (RNA) II (DNA directed) polypeptide A (220kD), polymerase (RNA) II (DNA directed) polypeptide A, 220kDa, RNA Pol 2, RNA Polymerase 2, RNA polymerase II subunit B1, RNA-directed RNA polymerase II subunit RPB1, RNAPII, RPB1, RPBh1, RpIILS, RPO2, RPOL2 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Xenopus species, Yeast, Zebrafish | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction