missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ RNA Marker Low

Product Code. 16002038
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16002038 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16002038 Supplier Abnova™ Supplier No. R0001.50ug
 Restricted Item
Estimated Shipment: 26-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

The RNA Marker Low consists of seven single-stranded RNAs.

  • The 20-base and 50-base RNA are synthesized by chemically (not phosphorylated).
  • The 100, 200, 300, 400 and 500 bases are synthesized by in vitro transcription.
  • The RNA Marker Low is suitable for determinating size of single-stranded RNAs in denaturing polyacrylamide gel electrophoresis.
  • The concentration of each RNA (20-500 base) in the marker is approximately 0.1 ug/ul.
  • It is useful for estimating of RNA amount.
  • The RNA Marker Low can be visualized by ethidium bromide staining.

Specifications

For Use With (Application) Electrophoresis
Quantity 50 μg
Storage Requirements Store at -80°C Repeated freeze/thaw cycles should be avoided
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.