missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ RNA Marker Low Easy

Product Code. 16012038
Change view
Click to view available options
Quantity:
125 μL
Unit Size:
125µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16012038 125 μL 125µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16012038 Supplier Abnova™ Supplier No. R0002.125uL
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

The RNA Marker Low Easy is supplied in a ready-to-use mixture of loading dye (containing formamide, EDTA sodium salt, bromphenol blue) and RNAs

  • It is prepared for denaturing polyacrylamide gel electrophoresis but not agarose gel elctrophoresis.
  • The RNA Marker Low Easy has seven single-stranded RNAs, 20, 50, 100, 200, 300, 400 and 500 bases.
  • The 20-base and 50-base RNA are synthesized by chemically (not phosphorylated), others are synthesized by in vitro transcription.
  • In 5 ul of the RNA Marker Low Easy, each RNA amount is approximately 100 ng.
  • It is useful for estimating RNA amount approximately.
  • The RNA Marker Low Easy can be visualized by UV light after ethidium bromide staining.

Specifications

For Use With (Application) Electrophoresis
Quantity 125 μL
Storage Requirements Store at -80°C Repeated freeze/thaw cycles should be avoided
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.