missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ RNA Marker High

Product Code. 16022038
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16022038 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16022038 Supplier Abnova™ Supplier No. R0003.50ug
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Suitable for determinating size of single-stranded RNAs in denaturing agarose gel electrophoresis

  • The RNA Marker High consists of nine single-stranded RNAs, 200, 500, 1,000, 1,500, 2,000, 3,000, 4,000, 5,000 and 8,000 bases, which are synthesized by in vitro transcription.
  • The RNA Marker High is suitable for determinating size of single-stranded RNAs in denaturing agarose gel electrophoresis.
  • The concentration of each RNA (200-8,000 base) in the marker is approximately 0.1 ug/ul.
  • It is useful for estimating of RNA amount.
  • The RNA Marker High can be visualized by UV light after ethidium bromide staining or exposure to film with end labeling.

Specifications

For Use With (Application) Electrophoresis
Quantity 50 μg
Storage Requirements Store at -80°C Repeated freeze/thaw cycles should be avoided
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.