missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ RNA Marker High Easy

Product Code. 16032038
Change view
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16032038 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16032038 Supplier Abnova™ Supplier No. R0004.25ug
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

For easy measurement of RNA size on agarose gel electrophoresis

  • The RNA Marker High Easy is manufactured for easy measurement of RNA size on agarose gel electrophoresis.
  • With RNA Marker High Easy, electrophorsis is possible on non-denaturing agarose gel as well as on denaturing agarose gel.
  • To use RNA Marker High Easy, just mix RNA loading buffer AG with formaldehyde solution (not included in this kit), and added the mixture to RNA Marker High Easy or RNA sample, then heat to load into wells of agarose gel.
  • The RNA loading buffer AG+ is a gel loading buffer prepared not only for denaturing agarose gel electrophoresis but also for non-contained denaturing agarose gel electrophoresis.
  • RNA Marker High Easy contains nine single-stranded RNAs, 500, 1,000, 1,500, 2,000, 3,000, 4,000, 5,000 and 8,000 bases.
  • Each RNA is 0.1 ug/ul approximately.
  • It is useful for estimating RNA amount approximately.
  • After electrophoresis, RNA bands on agarose gel can be visualized by UV light without further ethidium bromide staining.

Specifications

For Use With (Application) Electrophoresis
Quantity 25 μg
Storage Requirements Store at -80°C Repeated freeze/thaw cycles should be avoided
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.