missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNA Helicase A Antibody (S03-8A8), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19761-100ul
This item is not returnable.
View return policy
Description
RNA Helicase A Monoclonal antibody specifically detects RNA Helicase A in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| RNA Helicase A | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| S03-8A8 | |
| Unconjugated | |
| DDX9ATP-dependent RNA helicase A, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 9, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 9 (RNA helicase A, nuclear DNAhelicase II; leukophysin), DEAH (Asp-Glu-Ala-His) box polypeptide 9, DEAH box protein 9, EC 3.6.1, EC 3.6.1.15, EC 3.6.4.13, FLJ17406, leukophysin, LKP, NDH II, NDH2, NDHII, Nuclear DNA helicase II, RHA | |
| A synthetic peptide of human RNA Helicase A (Uniprot # Q08211) | |
| 100 μg | |
| Core ESC Like Genes, Stem Cell Markers | |
| 1660 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction