missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNA Helicase A Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-28915
This item is not returnable.
View return policy
Description
RNA Helicase A Polyclonal specifically detects RNA Helicase A in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| RNA Helicase A | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| DDX9ATP-dependent RNA helicase A, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 9, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 9 (RNA helicase A, nuclear DNAhelicase II; leukophysin), DEAH (Asp-Glu-Ala-His) box polypeptide 9, DEAH box protein 9, EC 3.6.1, EC 3.6.1.15, EC 3.6.4.13, FLJ17406, leukophysin, LKP, NDH II, NDH2, NDHII, Nuclear DNA helicase II, RHA | |
| Goat | |
| 141 kDa | |
| 0.1 mg | |
| Primary | |
| Human. Predicted cross-reactivity based on sequence identity: Mouse, Rat, Cow. | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:16000 | |
| Q08211 | |
| DHX9 | |
| Peptide with sequence TEGRNALIHKSSVNC corresponding to internal region according to NP_001348.2. | |
| Affinity Purified | |
| RUO | |
| 1660 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction