missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ RNA Helicase A Antibody (1D10), Novus Biologicals™

Product Code. 18345907 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18345907 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18345907 Supplier Novus Biologicals™ Supplier No. H00001660M03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

RNA Helicase A Monoclonal antibody specifically detects RNA Helicase A in Human samples. It is validated for ELISA,Western Blot,Sandwich ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNA Helicase A
Applications ELISA, Western Blot, Sandwich ELISA
Classification Monoclonal
Clone 1D10
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_001348
Gene Alias DDX9ATP-dependent RNA helicase A, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 9, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 9 (RNA helicase A, nuclear DNAhelicase II; leukophysin), DEAH (Asp-Glu-Ala-His) box polypeptide 9, DEAH box protein 9, EC 3.6.1, EC 3.6.1.15, EC 3.6.4.13, FLJ17406, leukophysin, LKP, NDH II, NDH2, NDHII, Nuclear DNA helicase II, RHA
Host Species Mouse
Immunogen DHX9 (NP_001348, 1 a.a. ∽ 90 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MGDVKNFLYAWCGKRKMTPSYEIRAVGNKNRQKFMCEVQVEGYNYTGMGNSTNKKDAQSNAARDFVNYLVRINEIKSEEVPAFGVASPPP
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 1660
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.