missing translation for 'onlineSavingsMsg'
Learn More

RNA binding motif protein, X-linked 2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18320210 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18320210 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18320210 Supplier Novus Biologicals Supplier No. NBP2834520.1ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RNA binding motif protein, X-linked 2 Polyclonal specifically detects RNA binding motif protein, X-linked 2 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNA binding motif protein, X-linked 2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 2% Sucrose
Gene Alias CGI-79,2810411A03Rik, RNA binding motif protein, X-linked 2, RNA-binding motif protein, X-linked 2
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the middle region of human RNA binding motif protein, X-linked 2
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51634
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.