missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ RN 9893 hydrochloride

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
RN 9893 hydrochloride is a potent and selective TRPV4 antagonist (IC50 values are 320, 420 and 660 nM for mouse, human and rat channels, respectively). Exhibits >15-fold selectivity for TRPV4 over TRPV1, TRPV3 and TRPM8, and a panel of 54 other common biological targets. Orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | N-[4-[[4-(1-Methylethyl)-1-piperazinyl]sulfonyl]phenyl]-2-nitro-4-(trifluoromethyl)benzamide hydrochloride |
| CAS | 2109450-40-8 |
| Quantity | 10 mg |
| Target | TRPV Antagonists |
| Molecular Formula | C21H23F3N4O5S.HCl |
| Formula Weight | 536.95 |
| Purity | >99% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction