missing translation for 'onlineSavingsMsg'
Learn More

RN 1747, Tocris Bioscience™

Catalog Number p-4816869
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
1024448-59-6
C17H18ClN3O4S
395.858
ZNLVYSJQUMALEO-UHFFFAOYSA-N
4-chloro-2-nitro-1-4-benzylpiperazinyl sulfonyl benzene, 1-benzyl-4-4-chloro-2-nitrobenzenesulfonyl piperazine, d0a5ly, rn hplc, 1-4-chloro-2-nitrophenyl sulfonyl-4-benzylpiperazine, 1-benzyl-4-4-chloro-2-nitrophenyl sulfonylpiperazine, 1-4-chloro-2-nitrophenyl sulfonyl-4-benzylpiperazine;, 1-benzyl-4-4-chloro-2-nitrobenzene sulfonyl piperazine, 1-4-chloro-2-nitrophenyl sulfonyl-4-phenylmethyl-piperazine
5068295
1-benzyl-4-(4-chloro-2-nitrophenyl)sulfonylpiperazine
C1CN(CCN1CC2=CC=CC=C2)S(=O)(=O)C3=C(C=C(C=C3)Cl)[N+](=O)[O-]
This item is not returnable. View return policy
1024448-59-6
C17H18ClN3O4S
395.858
ZNLVYSJQUMALEO-UHFFFAOYSA-N
4-chloro-2-nitro-1-4-benzylpiperazinyl sulfonyl benzene, 1-benzyl-4-4-chloro-2-nitrobenzenesulfonyl piperazine, d0a5ly, rn hplc, 1-4-chloro-2-nitrophenyl sulfonyl-4-benzylpiperazine, 1-benzyl-4-4-chloro-2-nitrophenyl sulfonylpiperazine, 1-4-chloro-2-nitrophenyl sulfonyl-4-benzylpiperazine;, 1-benzyl-4-4-chloro-2-nitrobenzene sulfonyl piperazine, 1-4-chloro-2-nitrophenyl sulfonyl-4-phenylmethyl-piperazine
5068295
1-benzyl-4-(4-chloro-2-nitrophenyl)sulfonylpiperazine
C1CN(CCN1CC2=CC=CC=C2)S(=O)(=O)C3=C(C=C(C=C3)Cl)[N+](=O)[O-]

Selective TRPV4 agonist

Selective TRPV4 agonist (EC50 values are 0.77, 4.0 and 4.1 μM for hTRPV4, mTRPV4 and rTRPV4 respectively). Displays selectivity over other TRP channels (EC50 values are 0.77, >30, >30 and >100 μM for TRPV4, TRPM8, TRPV3 and TRPV1 receptors respectively). Also antagonizes TRPM8 at relevant concentrations (IC50 = 4 μM).
Quantity 10 mg
Formula Weight 395.87
Percent Purity >98%
Chemical Name or Material RN 1747
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'