missing translation for 'onlineSavingsMsg'
Learn More

RN 1747, Tocris Bioscience™

Product Code. 18054492
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18054492 10 mg 10mg
18003912 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. Se returpolicy
Produktkod. 18054492 Leverantör Tocris Bioscience Leverantörsnummer 3745/10

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

Denna artikel kan inte returneras. Se returpolicy

Selective TRPV4 agonist

Selective TRPV4 agonist (EC50 values are 0.77, 4.0 and 4.1 μM for hTRPV4, mTRPV4 and rTRPV4 respectively). Displays selectivity over other TRP channels (EC50 values are 0.77, >30, >30 and >100 μM for TRPV4, TRPM8, TRPV3 and TRPV1 receptors respectively). Also antagonizes TRPM8 at relevant concentrations (IC50 = 4 μM).
TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 1024448-59-6
Molecular Formula C17H18ClN3O4S
Molecular Weight (g/mol) 395.858
InChI Key ZNLVYSJQUMALEO-UHFFFAOYSA-N
Synonym 4-chloro-2-nitro-1-4-benzylpiperazinyl sulfonyl benzene, 1-benzyl-4-4-chloro-2-nitrobenzenesulfonyl piperazine, d0a5ly, rn hplc, 1-4-chloro-2-nitrophenyl sulfonyl-4-benzylpiperazine, 1-benzyl-4-4-chloro-2-nitrophenyl sulfonylpiperazine, 1-4-chloro-2-nitrophenyl sulfonyl-4-benzylpiperazine;, 1-benzyl-4-4-chloro-2-nitrobenzene sulfonyl piperazine, 1-4-chloro-2-nitrophenyl sulfonyl-4-phenylmethyl-piperazine
PubChem CID 5068295
IUPAC Name 1-benzyl-4-(4-chloro-2-nitrophenyl)sulfonylpiperazine
SMILES C1CN(CCN1CC2=CC=CC=C2)S(=O)(=O)C3=C(C=C(C=C3)Cl)[N+](=O)[O-]

Specifikationer

Quantity 10 mg
Formula Weight 395.87
Percent Purity >98%
Chemical Name or Material RN 1747
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.