missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ RN 1734
GREENER_CHOICE

Product Code. 18791738
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
This item is not returnable. View return policy

Product Code. 18791738

Brand: Tocris Bioscience™ 3746/5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective TRPV4 antagonist

RN 1734 is a selective TRPV4 antagonist (IC50 values are 2.3, 3.2 and 5.9 μM for hTRPV4, rTRPV4 and mTRPV4 receptors respectively). Displays selectivity for TRPV4 over other TRP channels (IC50 values are 2.3, >30, >30 and >100 μM for TRPV4, TRPV3, TRPM8 and TRPV1 respectively). Blocks TRPV4-mediated arteriole vasodilation and increases myogenic tone ex vivo.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,4-Dichloro-N-isopropyl-N-(2-isopropylaminoethyl)benzenesulfonamide
CAS 946387-07-1
Quantity 5 mg
Molecular Formula C14H22Cl2N2O2S
Formula Weight Observed MW: 353.31
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.