missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ RN 1 dihydrochloride

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Potent lysine specific demethylase 1 (LSD1) inhibitor (IC50 = 70 nM). Exhibits selectivity for LSD1 over MAO-A and MAO-B (IC50 values are 0.51 and 2.79 μM for MAO-A and MAO-B, respectively). Impairs long-term, but not short-term memory in mice. Brain penetrant following systemic administration.
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 452.42 |
| Chemical Name or Material | RN 1 dihydrochloride |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction