missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Non-mammalian-based media composed of recombinant Protein A coupled with Sepharose 4 Fast Flow base matrix by reductive amination
- Coupling chemistry that minimizes leakage of protein A ligand
- Multi-point attachment of rProtein A by chemically-stable amide bonds
- High capacity and good pressure/flow characteristics provide for high throughput
- Matrix: 4% cross-linked agarose
- Average particle size: 90μm (d50V)
- Ligand: recombinant protein A (E. coli)
- pH Stability working range: 3 to 10
- pH Stability cleaning-in-place: 2 to 11
- Binding capacity/mL chromatography medium: >35mg human IgG/mL
- Pressure/Flow specification: base matrix 150 to 250cm/hr., 100kPa, XK 50/60 column
For therapeutic applications requiring extremely pure eluate fractions of antibodies with non-exposure of antibodies to human or animal derivatives during purification process.
Specifications
Specifications
| Quantity | 5 mL |
| Stability | Stable in aqueous Buffers commonly used in Protein A Chromatography |
| For Use With (Application) | For High-Purity Separation of Monoclonal and Polyclonal Antibodies |
| Storage Requirements | 2°C to 8°C |
| Binding Capacity | >Min. 22 mg Human IgG/mL |
| Product Type | rmp Protein A Sepharose Fast Flow Media |
| Flow Rate | 150 to 250 cm/Hoμr |
| Particle Size | 60 to 165 μm |
| Height (English) Bed | 9.84 in. |
| Height (Metric) Bed | 25 cm |
| Show More |
Safety and Handling
Product Identifier
- rmp Protein A Sepharose FF
- Warning
- Flammable liquid Category 3
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: ethanol
- UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?