missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RMND5A Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5110301

Product Code. 17108233

  • £440.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: ISLLMGGTTN QREALQYAKN FQPFALNHQK DIQVLMGSLV YLRQGIENSP YVHLLDANQW ADICDIFTRD A.

RMND5A is a protein coding gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RMND5A
Polyclonal
Unconjugated
RMND5A
1110007A06Rik; 44-kD protein coding for CTLH motif; AJ237586; AW125795; C-terminal to LisH motif, 44 kDa; CTLH; E3 ubiquitin-protein ligase RMND5A; E3 ubiquitin-protein transferase RMND5A; FLJ13910; GID complex subunit 2 homolog A; GID2; GID2A; MTA.D02.090; p44CTLH; Protein RMD5 homolog A; required for meiotic nuclear division 5 homolog A; required for meiotic nuclear division 5 homolog A (S. cerevisiae); RGD1309766; RMD5; RMND5A
Rabbit
Affinity Chromatography
RUO
312439, 64795, 68477
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
2.36 mg/mL
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Q80YQ8, Q9H871
RMND5A
Recombinant fusion protein containing a sequence corresponding to amino acids 200-270 of human RMND5A (NP_073617.1).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.