missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RMND1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA598968
This item is not returnable.
View return policy
Description
RMND1 Polyclonal Antibody for Western Blot, IHC (P), ELISA
The function of this protein remains unknown.
Specifications
| RMND1 | |
| Polyclonal | |
| Unconjugated | |
| RMND1 | |
| 0610042C05Rik; AA408137; AI462664; AW536662; bA351K16; bA351K16.3; C6orf96; COXPD11; Iag-1; implantation-associated gene-1; required for meiotic nuclear division 1 homolog; required for meiotic nuclear division 1 homolog (S. cerevisiae); required for meiotic nuclear division protein 1 homolog; RGD1309546; RMD1; Rmnd1 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 55005 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q9NWS8 | |
| RMND1 | |
| Recombinant Human Required for meiotic nuclear division protein 1 homolog protein (35-160AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction