missing translation for 'onlineSavingsMsg'
Learn More

RMND1 Antibody, Novus Biologicals™

Product Code. 18348181 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18348181 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18348181 Supplier Novus Biologicals Supplier No. NBP305256

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RMND1 Polyclonal specifically detects RMND1 in Human, Mouse, Rat samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RMND1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3. with 0.05% Proclin 300
Gene Alias bA351K16, bA351K16.3, C6orf96, chromosome 6 open reading frame 96, FLJ20627, MGC117362, MGC149570, MGC88260, required for meiotic nuclear division 1 homolog (S. cerevisiae), required for meiotic nuclear division protein 1 homolog, RMD1
Host Species Rabbit
Immunogen Recombinant protein of human RMND1.
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55005
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.