missing translation for 'onlineSavingsMsg'
Learn More

RMC-4550, MedChemExpress

Product Code. 16464858
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16464858 10 mg 10mg
16435646 100 mg 100mg
16431117 50 mg 50mg
16457607 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16464858 Supplier Medchem Express Supplier No. HY116009
 Restricted Item
Estimated Shipment: 28-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress RMC-4550 is a potent, selective and allosteric inhibitor of SHP2, with an IC50 of 0.583 nM.

RMC-4550 is a potent, selective and allosteric inhibitor of SHP2, with an IC50 of 0.583 nM.

Specifications

Chemical Name or Material RMC-4550
CAS 2172651-73-7
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C21H26Cl2N4O2
Quantity 10 mg
Solubility Information DMSO : ≥ 260 mg/mL (594.48 mM)
SMILES ClC1=C(C(C2=NC(CO)=C(N=C2C)N3CCC4(CC3)CO[C@H]([C@H]4N)C)=CC=C1)Cl
Molecular Weight (g/mol) 437.36
Formula Weight 437.36
Percent Purity 98.61%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.