missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RM30 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5118073
This item is not returnable.
View return policy
Description
RM30 Polyclonal Antibody for Western Blot, IHC (P), IP
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein. Alternative splicing results in multiple transcript variants. Pseudogenes corresponding to this gene are found on chromosomes 6p and 12p. Read-through transcription also exists between this gene and the neighboring upstream lipoyltransferase 1 (LIPT1) gene.
Specifications
| RM30 | |
| Polyclonal | |
| PBS with 0.03% ProClin 300; pH 7 | |
| Q8TCC3 | |
| MRPL30 | |
| E. coli-derived Human MRPL30 fragment. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| Unconjugated | |
| MRPL30 | |
| 2310001L22Rik; 39S ribosomal protein L28, mitochondrial; 39S ribosomal protein L30, mitochondrial; HSPC249; L28mt; L30MT; Mitochondrial large ribosomal subunit protein uL30m; mitochondrial ribosomal protein L30; MRPL28; MRP-L28; MRPL28M; MRPL30; MRP-L30; RPML28 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 51263 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction