missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RLN3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26075-1-AP-20UL
This item is not returnable.
View return policy
Description
Relaxin 3 is part of the Relaxin-like peptide family that plays an important neuropeptide signaling role. Relaxin 3 is a ligand for two structurally related G protein-coupled receptors, human Relaxin receptor 3 and Relaxin receptor 4. The greatest levels of Relaxin 3 expression occur in the brain, specifically in the pons/medulla, with highest levels in the pars ventromedialis of the dorsal tegmental nucleus and with lower levels in the hippocampus and olfactory regions. Significant expression is also observed in the spleen, thymus, lung and ovary. Relaxin 3 inhibits Forskolin-stimulated cAMP accumulation in a dose-dependent manner in cells that express Relaxin receptor 3. Acute and repeated administration of Relaxin 3 causes a decrease in levels of the plasma thyroid stimulating hormone, suggesting that Relaxin 3 may play a role in long-term control of food intake.Specifications
| RLN3 | |
| Polyclonal | |
| Unconjugated | |
| RLN3 | |
| H3, insl7, Insulin like peptide 7, Insulin like peptide INSL7, Prorelaxin H3, relaxin 3, RLN3, RXN3, ZINS4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 117579 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8WXF3 | |
| RLN3 | |
| RLN3 Fusion Protein Ag23382 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction