missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RLBP1L1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-55291
This item is not returnable.
View return policy
Description
RLBP1L1 Polyclonal specifically detects RLBP1L1 in Human samples. It is validated for Western Blot.
Specifications
| RLBP1L1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| Cellular retinaldehyde-binding protein-like, clavesin 1, clavesin-1, CRALBPLC6orf212L, FLJ37248, MGC34646, retinaldehyde binding protein 1-like 1, Retinaldehyde-binding protein 1-like 1, RLBP1L1 | |
| Rabbit | |
| 41 kDa | |
| 100 μL | |
| Cellular Markers, Cellular Signaling, Neuronal Cell Markers | |
| 157807 | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q8IUQ0 | |
| CLVS1 | |
| Synthetic peptides corresponding to RLBP1L1(retinaldehyde binding protein 1-like 1) The peptide sequence was selected from the N terminal of RLBP1L1. Peptide sequence NFKADDPGIKRALIDGFPGVLENRDHYGRKILLLFAANWDQSRNSFTDIL. | |
| Affinity purified | |
| RUO | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction