Learn More
Corning™ rLaminin-521, Human
Corning™ extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.
Brand:  Corning™ 354224
Additional Details : Weight : 0.01400kg
Product Code. 15940151
  • £295.00 / 500µg
Estimated Shipment: 26-09-2024
to see stock.
Description
Description
Corning™ extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.
Corning™ rLaminin-521 is the next generation pluripotent stem cell (PSC) surface that enables researchers to streamline culture workflow, lower contamination risk, and scale-up efficiently. Corning has partnered with BioLamina for the supply of recombinant human Laminin 521 (rLaminin-521), a biocompatible, full length Laminin that enables feeder-free, clump-free, single-cell passaging of PSCs without ROCK-inhibitor.
Specifications
Specifications
500 μg
Vial
Recombinant Protein
Recombinant
Product Suggestions