missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RKIP/PBP Antibody (003), CoraFluor™ 1, Novus Biologicals™
Description
RKIP/PBP Monoclonal antibody specifically detects RKIP/PBP in Human samples. It is validated for ELISA,Western Blot
Specifications
Specifications
| Antigen | RKIP/PBP |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 003 |
| Conjugate | CoraFluor 1 |
| Formulation | PBS |
| Gene Alias | HCNP, hippocampal cholinergic neurostimulating peptide, Neuropolypeptide h3, PBPPEBP-1, PEBPHCNPpp, phosphatidylethanolamine binding protein 1, phosphatidylethanolamine-binding protein 1, prostatic binding protein, Prostatic-binding protein, Raf kinase inhibitory protein, RKIPRaf kinase inhibitor protein |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human RKIP/PBP (Accession#: NP_002558.1; Met1-Lys187). |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?