missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RKIP/PBP Antibody (003), Alexa Fluor™ 350, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90295AF350
This item is not returnable.
View return policy
Description
RKIP/PBP Monoclonal specifically detects RKIP/PBP in Human samples. It is validated for Western Blot, ELISA.
Specifications
| RKIP/PBP | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 3 | |
| Western Blot, ELISA | |
| HCNP, hippocampal cholinergic neurostimulating peptide, Neuropolypeptide h3, PBPPEBP-1, PEBPHCNPpp, phosphatidylethanolamine binding protein 1, phosphatidylethanolamine-binding protein 1, prostatic binding protein, Prostatic-binding protein, Raf kinase inhibitory protein, RKIPRaf kinase inhibitor protein | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human RKIP/PBP (NP_002558.1; Met1-Lys187). | |
| 0.1 mL | |
| Cancer, Phospho Specific | |
| 5037 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction