missing translation for 'onlineSavingsMsg'
Learn More

RKI-1447, TRC

Product Code. 30426997
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30426997 5 mg 5mg
30424453 1 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30426997 Supplier Toronto Research Chemicals Supplier No. TRCR5500005MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1342278-01-6
Molecular Formula C16 H14 N4 O2 S
Molecular Weight (g/mol) 326.37
IUPAC Name 1-[(3-hydroxyphenyl)methyl]-3-(4-pyridin-4-yl-1,3-thiazol-2-yl)urea
SMILES Oc1cccc(CNC(=O)Nc2nc(cs2)c3ccncc3)c1

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C16H14N4O2S/c21-13-3-1-2-11(8-13)9-18-15(22)20-16-19-14(10-23-16)12-4-6-17-7-5-12/h1-8,10,21H,9H2,(H2,18,19,20,22)
Formula Weight 326.0837
Chemical Name or Material RKI-1447
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.