missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RKHD3 Antibody, DyLight 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81777V
This item is not returnable.
View return policy
Description
RKHD3 Polyclonal antibody specifically detects RKHD3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| RKHD3 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| DyLight 405 | |
| DKFZp434J0617, KIAA2009, mex-3 homolog B (C. elegans), ring finger and KH domain containing 3, RING finger and KH domain-containing protein 3, RING finger protein 195, RKHD3MGC117199, RNA-binding protein MEX3B, RNF195MEX-3B | |
| RKHD3 antibody was raised against a 16 amino acid synthetic peptide from near the center of human RKHD3. The immunogen is located within amino acids 180 - 230 of RKHD3. Amino Acid Squence: TYIVTPSRDKEPVFEV | |
| 0.1 mL | |
| Zinc Finger | |
| 84206 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction