missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RKHD1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA595905

Product Code. 16598741

  • £449.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: AGARRSSGAG TPRHSPTLPE PGGLRLELPL SRRGAPDPVG ALSWRPPQGP VSFPGGAAFS TATSLPSSPA AAACAPLDSG ASENSRKPPS ASSAPALARE C.

Rkhd1, also known as TINO or MEX3D is a member of a novel family of four homologous human MEX3 proteins each containing two heterogeneous nuclear ribonucleoprotein K homology (KH) domains and one carboxy-terminal RING finger module. MEX3 proteins, including Rkhd1, are phosphoproteins that bind RNA through their KH domains and shuttle between the nucleus and the cytoplasm via the CRM1 export pathway. These proteins are a novel family of evolutionarily conserved RNA-binding proteins, differentially recruited to P bodies and potentially involved in post-transcriptional regulatory mechanisms. Rkhd1 binds to the AU-rich element in the 3'-untranslated region of Bcl-2 mRNA and is thought to be a negative regulator of Bcl-2 expression. Rkhd3 and Rkhd4, but not Rkhd1, co-localize with both the hDcp1a decapping factor and Argonaute (Ago) proteins in processing bodies (P bodies), recently characterized as centers of mRNA turnover.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RKHD1
Polyclonal
Unconjugated
Mex3d
BC059858; bcl-2 ARE RNA binding protein; KIAA2031; MEX3; mex3 homolog D (C. elegans); mex-3 RNA binding family member D; Mex3d; MEX-3D; OK/SW-cl.4; ring finger (C3HC4 type) and KH domain containing 1; ring finger and KH domain containing 1; RING finger and KH domain-containing protein 1; RING finger protein 193; RKHD1; RNA-binding protein MEX3D; RNF193; TINO
Rabbit
Affinity Chromatography
RUO
237400
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
0.54 mg/mL
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Q3UE17
Mex3d
A synthetic peptide corresponding to a sequence within amino acids 500-600 of human MEX3D (NP_9760493).
100 μL
Primary
Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.