missing translation for 'onlineSavingsMsg'
Learn More

RIZ1 Antibody, DyLight 405, Novus Biologicals™

Product Code. 18369452 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18369452 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18369452 Supplier Novus Biologicals Supplier No. NBP299748V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RIZ1 Polyclonal specifically detects RIZ1 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RIZ1
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 405
Dilution ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias EC 2.1.1.43, GATA-3 binding protein G3B, GATA-3-binding protein G3B, KMT8PR domain-containing protein 2, Lysine N-methyltransferase 8, MTB-ZFHUMHOXY1, PR domain containing 2, with ZNF domain, PR domain zinc finger protein 2, retinoblastoma protein-binding zinc finger protein, Retinoblastoma protein-interacting zinc finger protein, RIZ1, RIZ2, RIZMTE-binding protein, Zinc finger protein RIZ, zinc-finger DNA-binding protein
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant RIZ1 (NP_036363.2; Met 1-Ala 200).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Chromatin Research
Primary or Secondary Primary
Gene ID (Entrez) 7799
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.