missing translation for 'onlineSavingsMsg'
Learn More

Rivastigmine Tartrate Salt, TRC

Product Code. 30136442
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136442 100 mg 100mg
30135450 10 mg 10mg
18073830 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136442 Supplier Toronto Research Chemicals Supplier No. TRCR541000100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 129101-54-8
Molecular Formula C14 H22 N2 O2 . C4 H6 O6
Molecular Weight (g/mol) 400.42
Synonym Rivastigmine Tartrate, Rivastigmine hydrogen tartrate, 3-[(1S)-1-(Dimethylamino)ethyl]phenyl ethyl(methyl)carbamate hydrogen (2R,3R)-2,3-dihydroxybutanedioate, Carbamic acid, N-ethyl-N-methyl-, 3-[(1S)-1-(dimethylamino)ethyl]phenyl ester, (2R,3R)-2,3-dihydroxybutanedioate (1:1)Carbamic acid, ethylmethyl-, 3-[(1S)-1-(dimethylamino)ethyl]phenyl ester, (2R,3R)-2,3-dihydroxybutanedioate (1:1) (9CI), Carbamic acid, ethylmethyl-, 3-[1-(dimethylamino)ethyl]phenyl ester, (S)-, [R-(R*,R*)]-2,3-dihydroxybutanedioate (1:1), ENA 713, Rivastigmine hydrogentartrate, Rivastigmine tartrate, SDZ-ENA 713, rivastigmine bitartrate
IUPAC Name (2R,3R)-2,3-dihydroxybutanedioic acid;[3-[(1S)-1-(dimethylamino)ethyl]phenyl] N-ethyl-N-methylcarbamate
SMILES CCN(C)C(=O)Oc1cccc(c1)[C@H](C)N(C)C.O[C@H]([C@@H](O)C(=O)O)C(=O)O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C14H22N2O2.C4H6O6/c1-6-16(5)14(17)18-13-9-7-8-12(10-13)11(2)15(3)4;5-1(3(7)8)2(6)4(9)10/h7-11H,6H2,1-5H3;1-2,5-6H,(H,7,8)(H,9,10)/t11-;1-,2-/m01/s1
Formula Weight 400.1846
Chemical Name or Material Rivastigmine hydrogen tartrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.