missing translation for 'onlineSavingsMsg'
Learn More

Rivastigmine tartrate, MedChemExpress

Product Code. 16485266
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
200 mg
Unit Size:
100mg
200mg
50mg
Product Code. Quantity unitSize
16485266 100 mg 100mg
16450687 50 mg 50mg
16431016 200 mg 200mg
3 options
This item is not returnable. View return policy

Product Code. 16485266

Brand: Medchem Express HY11017

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Rivastigmine tartrate (ENA 713; SDZ-ENA 713) is an orally active and potent cholinesterase (ChE) inhibitor and inhibits butyrylcholinesterase (BChE) and acetylcholinesteras (AChE) with IC50s of 0.037 μM, 4.15 μM, respectively.

Rivastigmine tartrate can pass the blood brain barrier (BBB). Rivastigmine tartrate is a parasympathomimetic or cholinergic agent used for the research of mild to moderate dementia of the Alzheimer's type and dementia due to Parkinson's disease.

Specifications

Chemical Name or Material Rivastigmine tartrate
CAS 129101-54-8
Color White
Purity Grade Notes Research
Molecular Formula C18H28N2O8
Quantity 100 mg
Synonym ENA 713 SDZ-ENA 713
Solubility Information H2O : 50 mg/mL (124.87 mM; Need ultrasonic)
SMILES O=C(OC1=CC=CC([C@@H](N(C)C)C)=C1)N(CC)C.O=C(O)[C@H](O)[C@@H](O)C(O)=O
Molecular Weight (g/mol) 400.42
Formula Weight 400.42
Percent Purity 99.71%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.