missing translation for 'onlineSavingsMsg'
Learn More

Rivastigmine, MedChemExpress

Product Code. 16497256
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
50mg
This item is not returnable. View return policy

Product Code. 16497256

Brand: Medchem Express HY17368

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Rivastigmine (S-Rivastigmine) is an orally active and potent cholinesterase (ChE) inhibitor and inhibits butyrylcholinesterase (BChE) and acetylcholinesteras (AChE) with IC50s of 0.037 μM , 4.15 μM, respectively.

Rivastigmine can pass the blood brain barrier (BBB). Rivastigmine is a parasympathomimetic or cholinergic agent used for the research of mild to moderate dementia of the Alzheimer's type and dementia due to Parkinson's disease.

Specifications

Chemical Name or Material Rivastigmine
CAS 123441-03-2
Color Colorless
Purity Grade Notes Research
Molecular Formula C14H22N2O2
Quantity 100 mg
Synonym S-Rivastigmine
Solubility Information DMSO : ≥ 50 mg/mL (199.73 mM)
SMILES O=C(N(C)CC)OC1=CC=CC([C@H](C)N(C)C)=C1
Molecular Weight (g/mol) 250.34
Formula Weight 250.34
Percent Purity 98.75%
Grade Research
Physical Form Liquid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.