missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rivastigmine Hydrogen Tartrate, LoGiCal
Specifications
Specifications
| Chemical Name or Material | Rivastigmine hydrogen tartrate |
| CAS | 129101-54-8 |
| Analyte or Component Names | Rivastigmine hydrogen tartrate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C14 H22 N2 O2 . C4 H6 O6 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Rivastigmine Tartrate, Rivastigmine hydrogen tartrate, 3-[(1S)-1-(Dimethylamino)ethyl]phenyl ethyl(methyl)carbamate hydrogen (2R,3R)-2,3-dihydroxybutanedioate, Carbamic acid, N-ethyl-N-methyl-, 3-[(1S)-1-(dimethylamino)ethyl]phenyl ester, (2R,3R)-2,3-dihydroxybutanedioate (1:1)Carbamic acid, ethylmethyl-, 3-[(1S)-1-(dimethylamino)ethyl]phenyl ester, (2R,3R)-2,3-dihydroxybutanedioate (1:1) (9CI), Carbamic acid, ethylmethyl-, 3-[1-(dimethylamino)ethyl]phenyl ester, (S)-, [R-(R*,R*)]-2,3-dihydroxybutanedioate (1:1), ENA 713, Rivastigmine hydrogentartrate, Rivastigmine tartrate, SDZ-ENA 713, rivastigmine bitartrate |
| InChI Formula | InChI=1S/C14H22N2O2.C4H6O6/c1-6-16(5)14(17)18-13-9-7-8-12(10-13)11(2)15(3)4;5-1(3(7)8)2(6)4(9)10/h7-11H,6H2,1-5H3;1-2,5-6H,(H,7,8)(H,9,10)/t11-;1-,2-/m01/s1 |
| Solution Type | Neat |
| SMILES | CCN(C)C(=O)Oc1cccc(c1)[C@H](C)N(C)C.O[C@H]([C@@H](O)C(=O)O)C(=O)O |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?