missing translation for 'onlineSavingsMsg'
Learn More

Rivastigmine Hydrogen Tartrate Impurity B (as Racemate Hydrochloride), Mikromol™

Product Code. 30457884
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30457884 4 x 25 mg 100mg
30052624 25 mg 25mg
30049292 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30457884 Supplier Mikromol Supplier No. MM1497.0600254AMP
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Rivastigmine Hydrogen Tartrate Impurity B (as Racemate Hydrochloride), Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-[(1RS)-1-(Dimethylamino)ethyl]phenyl Dimethylcarbamate Hydrochloride
CAS 105601-13-6
Analyte or Component Names 3-[(1RS)-1-(Dimethylamino)ethyl]phenyl Dimethylcarbamate Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C13 H20 N2 O2 . Cl H
Certifications/Compliance ISO 9001
Synonym Rivastigmine Hydrogen Tartrate Imp. B (EP), Rivastigmine Imp. B (EP), Rivastigmine USP Related Compound B, Rivastigmine USP RC B, Carbamic acid, dimethyl-, 3-[1-(dimethylamino)ethyl]phenyl ester, monohydrochloride (9CI), 3-[(1RS)-1-(Dimethylamino)ethyl]phenyl Dimethylcarbamate Hydrochloride, 3-[(1RS)-1-(Dimethylamino)ethyl]phenyl N,N-Dimethylcarbamate Hydrochloride, Rivastigmine Related Compound B as Hydrochloride, Rivastigmine Hydrogen Tartrate Impurity B as Racemate Hydrochloride, Rivastigmine Impurity B as Racemate Hydrochloride
InChI Formula InChI=1S/C13H20N2O2.ClH/c1-10(14(2)3)11-7-6-8-12(9-11)17-13(16)15(4)5;/h6-10H,1-5H3;1H
Solution Type Neat
SMILES Cl.CC(N(C)C)c1cccc(OC(=O)N(C)C)c1
IUPAC Name [3-[1-(dimethylamino)ethyl]phenyl] N,N-dimethylcarbamate;hydrochloride
Molecular Weight (g/mol) 272.77
Quantity 4 x 25 mg
Formula Weight 272.1292 g/mol
Show More Show Less
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.