missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rivastigmine Hydrogen Tartrate Impurity B (as Racemate Hydrochloride), Mikromol™
Description
Rivastigmine Hydrogen Tartrate Impurity B (as Racemate Hydrochloride)
Specifications
Specifications
| Chemical Name or Material | 3-[(1RS)-1-(Dimethylamino)ethyl]phenyl Dimethylcarbamate Hydrochloride |
| CAS | 105601-13-6 |
| Analyte or Component Names | 3-[(1RS)-1-(Dimethylamino)ethyl]phenyl Dimethylcarbamate Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C13 H20 N2 O2 . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | Rivastigmine Hydrogen Tartrate Imp. B (EP), Rivastigmine Imp. B (EP), Rivastigmine USP Related Compound B, Rivastigmine USP RC B, Carbamic acid, dimethyl-, 3-[1-(dimethylamino)ethyl]phenyl ester, monohydrochloride (9CI), 3-[(1RS)-1-(Dimethylamino)ethyl]phenyl Dimethylcarbamate Hydrochloride, 3-[(1RS)-1-(Dimethylamino)ethyl]phenyl N,N-Dimethylcarbamate Hydrochloride, Rivastigmine Related Compound B as Hydrochloride, Rivastigmine Hydrogen Tartrate Impurity B as Racemate Hydrochloride, Rivastigmine Impurity B as Racemate Hydrochloride |
| InChI Formula | InChI=1S/C13H20N2O2.ClH/c1-10(14(2)3)11-7-6-8-12(9-11)17-13(16)15(4)5;/h6-10H,1-5H3;1H |
| SMILES | Cl.CC(N(C)C)c1cccc(OC(=O)N(C)C)c1 |
| IUPAC Name | [3-[1-(dimethylamino)ethyl]phenyl] N,N-dimethylcarbamate;hydrochloride |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?