missing translation for 'onlineSavingsMsg'
Learn More

Rivaroxaban, MedChemExpress

Product Code. 16474047
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16474047 50 mg 50mg
16448138 10 mg 10mg
16420598 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16474047 Supplier Medchem Express Supplier No. HY50903
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Rivaroxaban (BAY 59-7939) is a highly potent, selective and direct Factor Xa (FXa) inhibitor, achieving a strong gain in anti-FXa potency (IC50 0.7 nM; Ki 0.4 nM).

Rivaroxaban (BAY 59-7939) is a highly potent, selective and direct Factor Xa (FXa) inhibitor, achieving a strong gain in anti-FXa potency (IC50 0.7 nM

Specifications

Chemical Name or Material Rivaroxaban
Physical Form Solid
CAS 366789-02-8
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C19H18ClN3O5S
For Use With (Application) COVID-19-immunoregulation
Synonym BAY 59-7939
Solubility Information DMSO : 50 mg/mL (114.71 mM; Need ultrasonic)
SMILES O=C(N(C1=CC=C(N2C(COCC2)=O)C=C1)C3)O[C@H]3CNC(C4=CC=C(S4)Cl)=O
Molecular Weight (g/mol) 435.88
Formula Weight 435.88
Percent Purity 99.67%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.