missing translation for 'onlineSavingsMsg'
Learn More

Rivanicline hemioxalate, MedChemExpress

Product Code. 16435568
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16435568

Brand: Medchem Express HY13225B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Rivanicline hemioxalate (RJR-2403 hemioxalate; (E)-Metanicotine hemioxalate) is a neuronal nicotinic receptor agonist, showing high selectivity for the α4β2 subtype (Ki=26 nM); > 1,000 fold selectivity than α7 receptors(Ki= 3.6 μM).

Rivanicline hemioxalate (RJR-2403 hemioxalate

Specifications

Chemical Name or Material Rivanicline hemioxalate
Physical Form Solid
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C11H15N2O2
Synonym RJR-2403 hemioxalate(E)-Metanicotine hemioxalate
Solubility Information DMSO : 50 mg/mL (241.28 mM; Need ultrasonic)
SMILES CNCC/C=C/C1=CC=CN=C1.OC(C(O)=O)=O.[0.5]
Molecular Weight (g/mol) 207.23
Formula Weight 207.23
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.