missing translation for 'onlineSavingsMsg'
Learn More

Ritonavir, MedChemExpress

Product Code. 16468248
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
500 mg
Unit Size:
100mg
10mg
500mg
50mg
This item is not returnable. View return policy

Product Code. 16468248

Brand: Medchem Express HY90001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ritonavir (ABT 538) is an inhibitor of HIV protease used to treat HIV infection and AIDS. Ritonavir is also a SARS-CoV 3CLpro inhibitor with an IC50 of 1.61 μM.

Ritonavir (ABT 538) is an inhibitor of HIV protease used to treat HIV infection and AIDS. Ritonavir is also a SARS-CoV 3CLpro inhibitor with an IC50 of 1.61 μM.

Specifications

Chemical Name or Material Ritonavir
Physical Form Powder
CAS 155213-67-5
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C37H48N6O5S2
For Use With (Application) COVID-19-anti-virus
Synonym ABT 538 RTV
Solubility Information DMSO : 25 mg/mL (34.68 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(N[C@@H](CC1=CC=CC=C1)[C@H](C[C@H](CC2=CC=CC=C2)NC([C@H](C(C)C)NC(N(CC3=CSC(C(C)C)=N3)C)=O)=O)O)OCC4=CN=CS4
Molecular Weight (g/mol) 720.94
Formula Weight 720.94
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.