missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Ritonavir
GREENER_CHOICE

Product Code. 18766919
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
This item is not returnable. View return policy

Product Code. 18766919

Brand: Tocris Bioscience™ 5856/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

HIV-1 and HIV-2 protease inhibitor

Ritonavir is a HIV-1 and HIV-2 protease inhibitor (EC50 values are 0.022-0.13 and 0.16 μM, respectively). Blocks the metabolism of protease inhibitors by liver enzyme cytochrome P450-3A4 (CYP3A4). Orally bioavailable. When used in combination with lopinavir, improves outcome in an animal model of MERS-CoV infection.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Thiazolylmethyl (3 S,4 S,6 S,9 S)-4-hydroxy-12-methyl-9-(1-methylethyl)-13-[2-(1-methylethyl)-4-thiazolyl]-8,11-dioxo-3,6-bis(phenylmethyl)-2,7,10,12-tetraazatridecanoate
CAS 155213-67-5
Synonym ABT-538, A-84538, Abbot 84538
Quantity 50 mg
Target HIV Protease Inhibitors
Molecular Formula C37H48N6O5S2
Formula Weight 720.94
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.