missing translation for 'onlineSavingsMsg'
Learn More

Ritter Plastic™ Ritips™ Sterile Dispenser Tips

Product Code. 16690302
Click to view available options
Volume (Metric):
0.05 mL
0.5 mL
1.25 mL
2.5 mL
5.0 mL
12.5 mL
25.0 mL
50.0 mL
Quantity:
100/Pk.
25/Pk.
Unit Size:
Pack of 1600
Pack of 400
This item is not returnable. View return policy

Product Code. 16690302

Brand: Ritter Plastic™ 400100010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Used for the repetitive distribution of volumes between 0.05 and 50 mL.

  • Uses direct displacement method
  • Works well for infectious and volatile liquids
  • Fluid never in touch with the instrument to avoid contamination
  • Automated individual tests of each single tip guarantee the tightness with water, which is documented in lot-specific records
  • Black graduation marks
  • Manufactured from polypropylene
  • Suitable for a wide range of pipettors
  • For sensitive applications or analysis of body fluids

Specifications

Volume (Metric) 1.25 mL
Sterility Sterile
For Use With (Application) Sensitive applications, body fluid analysis
Quantity 100/Pk.
Product Type Pipette Tip
For Use With (Equipment) Wide range of pipettors
Graduated Yes
Product Line RiTips
Material Polypropylene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.