missing translation for 'onlineSavingsMsg'
Learn More

RITA, MedChemExpress

Product Code. 16475678
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16475678 10 mg 10mg
16441847 50 mg 50mg
16408377 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16475678 Supplier Medchem Express Supplier No. HY13424
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress RITA is an inhibitor of p53-HDM-2 interaction, binds to p53dN, with a Kd of 1.5 nM, and also induces DNA-DNA cross-links.

RITA is an inhibitor of p53-HDM-2 interaction, binds to p53dN, with a Kd of 1.5 nM, and also induces DNA-DNA cross-links.

Specifications

Chemical Name or Material RITA
Physical Form Solid
CAS 213261-59-7
Color Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C14H12O3S2
For Use With (Application) Cancer-programmed cell death
Synonym NSC 652287
Solubility Information DMSO : ≥ 100 mg/mL (342.03 mM)
SMILES OCC1=CC=C(C2=CC=C(C3=CC=C(CO)S3)O2)S1
Molecular Weight (g/mol) 292.37
Formula Weight 292.37
Percent Purity 99.45%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.