missing translation for 'onlineSavingsMsg'
Learn More

Rit2 Antibody (OTI4A6), DyLight 594, Novus Biologicals™

Product Code. 18413475 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18413475 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18413475 Supplier Novus Biologicals Supplier No. NBP273914DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Rit2 Monoclonal antibody specifically detects Rit2 in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rit2
Applications Western Blot
Classification Monoclonal
Clone OTI4A6
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias GTP-binding protein Rit2, GTP-binding protein Roc2, Ras-like protein expressed in neurons, Ras-like without CAAX 2, Ras-like without CAAX protein 2, RIBA, Ric (Drosophila)-like, expressed in neurons, Ric-like, expressed in neurons, RIN, ROC2
Host Species Mouse
Immunogen Full length recombinant protein of human Rit2 (NP_002921) produced in HEK293T cells.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6014
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.