missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rit2 Antibody (OTI3F4), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-03062
This item is not returnable.
View return policy
Description
Rit2 Monoclonal specifically detects Rit2 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Rit2 | |
| Monoclonal | |
| 0.69 mg/mL | |
| Western Blot 1:500, Flow Cytometry 1:100, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:150 | |
| RIT2 | |
| Full length human recombinant protein of human RIT2 (NP_002921) produced in HEK293T cell. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Canine, Primate, Monkey | |
| IgG2a |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| OTI3F4 | |
| Unconjugated | |
| GTP-binding protein Rit2, GTP-binding protein Roc2, Ras-like protein expressed in neurons, Ras-like without CAAX 2, Ras-like without CAAX protein 2, RIBA, Ric (Drosophila)-like, expressed in neurons, Ric-like, expressed in neurons, RIN, ROC2 | |
| Mouse | |
| 24.5 kDa | |
| 0.1 mL | |
| Signal Transduction | |
| 6014 | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction