missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RIT1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5111371
This item is not returnable.
View return policy
Description
Immunogen sequence: AYRYYIDDVF HALVREIRRK EKEAVLAMEK KSKPKNSVWK RLKSPFRKKK DSVT Highest antigen sequence indentity to the following orthologs: Mouse - 93%, Rat - 91%.
Rit and its neuron-specific homologue, Rin, define a recently discovered subfamily of Ras-related GTPases. Rit and Rin are membrane-associated in spite of the fact that they lack a CAAX box or similar C-terminal lipidation motif. Rit and Rin display 64% amino acid sequence identity and share a unique nine amino acid effector domain (DPTIEDAYK) that is 100% conserved between the murine and human proteins. Although the effector domain sequences of Rit and Rin are very similar to that of Ras, Rit and Rin have been shown to interact with the known Ras-binding proteins RalGDS, Rlf and AF-6, but not the Raf kinases, RIN1 or the p110 subunit of PI3 kinase. For this reason, it has been suggested that Rit and Rin may play important roles in the regulation of signaling pathways distinct from those controlled by Ras.
Specifica
| RIT1 | |
| Polyclonal | |
| Unconjugated | |
| RIT1 | |
| GTP-binding protein Rit1; GTP-binding protein Roc1; MGC125864; MGC125865; NS8; Ras like without CAAX 1; ras-like protein expressed in many tissues; Ras-like without CAAX 1; ras-like without CAAX protein 1; RGD1559874; RIBB; Ric-like, expressed in many tissues; RIT; RIT1; ROC1; si:ch211-93a2.3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6016 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q92963 | |
| RIT1 | |
| Recombinant Protein corresonding to Human RIT1. Recombinant protein control fragment (Product #RP-101351). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto