missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RIT1 Monoclonal Antibody (456201)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561475
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Rit and its neuron-specific homologue, Rin, define a recently discovered subfamily of Ras-related GTPases. Rit and Rin are membrane-associated in spite of the fact that they lack a CAAX box or similar C-terminal lipidation motif. Rit and Rin display 64% amino acid sequence identity and share a unique nine amino acid effector domain (DPTIEDAYK) that is 100% conserved between the murine and human proteins. Although the effector domain sequences of Rit and Rin are very similar to that of Ras, Rit and Rin have been shown to interact with the known Ras-binding proteins RalGDS, Rlf and AF-6, but not the Raf kinases, RIN1 or the p110 subunit of PI3 kinase. For this reason, it has been suggested that Rit and Rin may play important roles in the regulation of signaling pathways distinct from those controlled by Ras.
Specifications
| RIT1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q92963 | |
| RIT1 | |
| E. coli-derived recombinant human Rit1, Met1-Thr219. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| 456201 | |
| Unconjugated | |
| RIT1 | |
| GTP-binding protein Rit1; GTP-binding protein Roc1; MGC125864; MGC125865; NS8; Ras like without CAAX 1; ras-like protein expressed in many tissues; Ras-like without CAAX 1; ras-like without CAAX protein 1; RGD1559874; RIBB; Ric-like, expressed in many tissues; RIT; RIT1; ROC1; si:ch211-93a2.3 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 6016 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction