missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rit1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-20169
This item is not returnable.
View return policy
Description
Rit1 Polyclonal specifically detects Rit1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Rit1 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| GTP-binding protein Rit1, GTP-binding protein Roc1, MGC125864, MGC125865, Ras-like protein expressed in many tissues, Ras-like without CAAX 1, Ras-like without CAAX protein 1, RIBBRIT, Ric-like, expressed in many tissues, RIT, ROC1Ric (Drosophila)-like, expressed in many tissues | |
| Rabbit | |
| 25 kDa | |
| 0.1 mL | |
| Signal Transduction, Vision | |
| 6016 | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 0.75mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| Q92963 | |
| RIT1 | |
| Recombinant protein encompassing a sequence within the center region of human RIT1. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human. Species reactivity based on sequence identity: Rhesus Monkey (100%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction