missing translation for 'onlineSavingsMsg'
Learn More

Risperidone EP Impurity J, TRC

Product Code. 17934795
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
Unit Size:
10mg
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17934795 10 mg 10mg
17911435 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17934795 Supplier Toronto Research Chemicals Supplier No. TRCF59159510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1346606-24-3
Molecular Formula C35 H40 F2 N6 O3
Molecular Weight (g/mol) 630.7273
Synonym 3-[2-[4-[(EZ)-[4-Fluoro-2-[4-(6-fluoro-1,2-benzisoxazol-3-yl)piperidin-1-yl]phenyl](hydroxyimino)methyl]piperidin-1-yl]ethyl]-2-methyl-6,7,8,9-tetrahydro-4H-pyrido[1,2-a]pyrimidin-4-one, Risperidone Imp. J (EP) as (E/Z)-mixture, 3-[2-[4-[(Z)-(4-Fluoro-2-[4-(6-fluoro-1,2-benzisoxazol-3-yl)piperidin-1-yl)phenyl](hydroxyimino)methyl]piperidin-1-yl]ethyl]-2-methyl-6,7,8,9-tetrahydro-4H-pyrido[1,2-a]pyrimidin-4-one, (Risperidone Impurity, Mixture of E/Z isomers)
IUPAC Name 3-[2-[4-[C-[4-fluoro-2-[4-(6-fluoro-1,2-benzoxazol-3-yl)piperidin-1-yl]phenyl]-N-hydroxycarbonimidoyl]piperidin-1-yl]ethyl]-2-methyl-6,7,8,9-tetrahydropyrido[1,2-a]pyrimidin-4-one
SMILES CC1=C(CCN2CCC(CC2)C(=NO)c3ccc(F)cc3N4CCC(CC4)c5noc6cc(F)ccc56)C(=O)N7CCCCC7=N1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C35H40F2N6O3/c1-22-27(35(44)43-14-3-2-4-32(43)38-22)13-17-41-15-9-23(10-16-41)33(39-45)28-7-5-25(36)20-30(28)42-18-11-24(12-19-42)34-29-8-6-26(37)21-31(29)46-40-34/h5-8,20-21,23-24,45H,2-4,9-19H2,1H3
Formula Weight 630.313
Chemical Name or Material Risperidone EP Impurity J
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.