missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Positioning Base
Description
- Base riser
- Available in 30 and 40 cm sizes
Specifications
Specifications
| Product Type | Riser Bases |
| Height (Metric) | 30.5 cm |
| For Use With (Equipment) | Innova™ 44/44R Stackable Incubator Shakers |
| Packaging Quantity | 1 Ea. |
| Size | Medium |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?