missing translation for 'onlineSavingsMsg'
Learn More

Risedronic acid, MedChemExpress

Product Code. 16468136
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16468136

Brand: Medchem Express HYB0148

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Risedronic acid (Risedronate ) is a pyridinyl biphosphonate which inhibits osteoclast-mediated bone resorption.

Risedronic acid (Risedronate ) is a pyridinyl biphosphonate which inhibits osteoclast-mediated bone resorption.

Specifications

Chemical Name or Material Risedronic acid
Physical Form Solid
CAS 105462-24-6
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C7H11NO7P2
Synonym Risedronate
Solubility Information 0.1 M NaOH : 11 mg/mL (38.85 mM; ultrasonic and adjust pH to 7 with NaOH) ∣H2O : 0.67 mg/mL (2.37 mM; Need ultrasonic)
SMILES OC(P(O)(O)=O)(P(O)(O)=O)CC1=CN=CC=C1
Molecular Weight (g/mol) 283.11
Formula Weight 283.11
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.